EMAIL THIS PAGE TO A FRIEND
Polish journal of pharmacology and pharmacy
Craviten hydrolysis in plasma in vitro.
PMID 7322949
Abstract
Craviten hydrolysis in vitro was studied in human, rat and canine plasma and erythrocytes. Craviten was degraded during the incubation with plasma and the hydrolysis was inhibited by trimethoxybenzoic acid, a metabolite of Craviten. Erythrocyte enzymes do not participate in the hydrolysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T69000
3,4,5-Trimethoxybenzoic acid, ReagentPlus®, 99%
C10H12O5