EMAIL THIS PAGE TO A FRIEND
Acta embryologiae et morphologiae experimentalis ("Halocynthia" Association")
Teratogenic activity of organophosphate pesticide in chick embryos.
PMID 7324789
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45305
Dicrotophos, PESTANAL®, analytical standard
C8H16NO5P