EMAIL THIS PAGE TO A FRIEND
Archivos de farmacologia y toxicologia
[Synergistic action of sodium thiophencarboxylate and bromhexine in chronic bronchitis in rats].
PMID 7325708
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T32603
2-Thiophenecarboxylic acid, ReagentPlus®, 99%
C5H4O2S