EMAIL THIS PAGE TO A FRIEND
Food and cosmetics toxicology
Long-term toxicity study of ethyl methylphenylglycidate (strawberry aldehyde) in the rat.
PMID 7327470
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W244406
Ethyl 3-methyl-3-phenylglycidate, ≥98%, FCC, FG
C12H14O3