EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Effects of 1-methyl cyclohexane carboxylic acid (CCA) on cellular energetics in neuroblastoma cells.
PMID 7332572
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
142824
1-Methyl-1-cyclohexanecarboxylic acid, 99%
C8H14O2