EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Characterization of fluorinated metabolites by a gas chromatographic-helium microwave plasma detector--the biotransformation of 1H, 1H, 2H, 2H-perfluorodecanol to perfluorooctanoate.
PMID 7337230
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
532789
1H,1H,2H,2H-Perfluoro-1-decanol, 97%
C10H5F17O