EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
Glycerol conformation and molecular packing of membrane lipids. The crystal structure of 2,3-dilauroyl-D-glycerol.
PMID 7338925
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9758
Dilaurin Mixed Isomers, ≥99%
C27H52O5