EMAIL THIS PAGE TO A FRIEND
Toxicological European research. Recherche europeenne en toxicologie
[Determination of low levels of diethylcarbamoyl chloride in air. First results after inhalation in rat (author's transl)].
PMID 7340002
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D91403
Diethylcarbamoyl chloride, 97%
C5H10ClNO