EMAIL THIS PAGE TO A FRIEND
South African medical journal = Suid-Afrikaanse tydskrif vir geneeskunde
The association between measured and clinically assessed amniotic fluid volume and fetal growth and nutrition.
PMID 734583
Abstract
A close correlation was found between amniotic fluid volume measured by using para-aminohippuric acid and the clinical assessment of volume. No correlation was found between measured amniotic fluid volume and fetal weight, length, or weight for gestational age.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1422
p-Aminohippuric acid, ≥99%
C9H10N2O3
08090
p-Aminohippuric acid, ≥98.0% (T)
C9H10N2O3