EMAIL THIS PAGE TO A FRIEND
Biochemistry
Studies on synthetic peptides that bind to fibrinogen and prevent fibrin polymerization. Structural requirements, number of binding sites, and species differences.
PMID 7356959
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1895
Gly-Pro-Arg-Pro, ≥95% (HPLC)
C18H31N7O5