EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Antiamebic amidines and sulfonamides of 5- and 6-amino-2,3-bis(4-alkyl-1-piperazinyl)quinoxalines.
PMID 7359534
Abstract
A series of amidines and sulfonamides of 5- and 6-amino-2,3-bis(4-alkyl-1-piperazinyl)quinoxalines was synthesized and tested against cecal and hepatic forms of Entamoeba histolytica infections in rats and hamsters, respectively. Four compounds (5, 6, 8, and 9) were found to have acceptable activity against infections in both species but were too toxic to be considered for use in man.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
141674
1-n-Propylpiperazine dihydrobromide, 96%
C7H16N2 · 2HBr