EMAIL THIS PAGE TO A FRIEND
Andrologia
[Free fatty acid composition of human semen in different clinical diagnoses].
PMID 736283
Abstract
The composition of free fatty acids of human semen from different clinical diagnosis was determined by gas liquid chromatography. The major components were palmitic, stearic, myristic and oleic acid. The analyses indicate that the percentage composition of palmitic acid was distinctly lower in the group with azoospermia.
Related Materials