EMAIL THIS PAGE TO A FRIEND
Psychiatria polska
[Possibility of occurrence of psychoses after pridinol treatment].
PMID 7367525
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4419
Pridinol methanesulfonate salt, analytical standard
C20H25NO · CH4O3S