EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
High-pressure liquid chromatographic analysis of pramoxine hydrochloride in high lipoid aerosol foam dosage form.
PMID 7373553
Abstract
A rapid and quantitative method for the determination of pramoxine hydrochloride by high-pressure liquid chromatography is presented. The drug is extracted as the salt from a preparation with a high lipoid composition by partitioning it to the aqueous phase of an ether-methanol-water-acetic acid system. The extract is chromatographed on an octadecylsilane bonded packing with a methanol-water-acetic acid-methanesulfonic acid mobile phase. The time required for each separation is approximately 6 min. Analytical recoveries of 100.4 +/- 1.5% were obtained.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9422
Pramoxine hydrochloride, analytical standard, ≥98%
C17H27NO3 · HCl