EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Establishment of the maximum permissible concentration of amiben in the water of reservoirs].
PMID 7380279
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
152668
3-Amino-2,5-dichlorobenzoic acid, 95%
C7H5Cl2NO2
33392
Chloramben, PESTANAL®, analytical standard
C7H5Cl2NO2