EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Self-catalyzed inactivation of hepatic cytochrome P-450 by ethynyl substrates.
PMID 7380828
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129216
Cyclohexylacetylene, 98%
C8H12