EMAIL THIS PAGE TO A FRIEND
Clinical toxicology
Carcinogenicity of Chlorobenzilate in mice, rats, and dogs.
PMID 7389283
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45376
Chlorobenzilate, PESTANAL®, analytical standard
C16H14Cl2O3