EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
2-Cyano-N-[(ethylamino)carbonyl]-2-(methoxyimino)acetamide, a new fungicide.
PMID 7391384
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34326
Cymoxanil, PESTANAL®, analytical standard
C7H10N4O3