EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Aporphines. 30. (-)-N(2-chloroethyl)-10,11-dihydroxynor-aporphine (chloroethylnorapomorphine), a novel irreversible dopamine receptor antagonist.
PMID 7392026
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D007
R(−)-Chloroethylnorapomorphine hydrochloride, analytical standard
C18H18ClNO2 · HCl