EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Further studies of effects of vehicles and elicitation concentration in experimental contact sensitization testing in humans.
PMID 7398265
Abstract
Further confirmation of the effects of vehicles and elicitation concentration in experimental contact sensitization testing with fragrance ingredients is reported. A dose-response relation was seen when sensitized human subjects were challenged with dihydrocoumarin, alantroot oil and diethylmalleate. Furthermore, alcohol was shown to be a more effective vehicle than petrolatum, when cinnamon bark oil, vetiver acetate and diethylmalleate were used in predictive tests. The relation of these findings to risk-benefit judgments is discussed briefly.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D104809
Dihydrocoumarin, 99%
C9H8O2
W238104
Dihydrocoumarin, ≥99%, FCC, FG
C9H8O2