EMAIL THIS PAGE TO A FRIEND
American Industrial Hygiene Association journal
Chlordane sampling efficiency with Chromosorb 102 and ethylene glycol.
PMID 7405829
Abstract
An organic polymeric solid, Chromosorb 102, has been reported to collect various pesticides from air. In this study the collection efficiency of Chromosorb 102 for chlordane was evaluated. Preparation of sampling tubes containing this medium for field sampling is also discussed.