EMAIL THIS PAGE TO A FRIEND
Hoppe-Seyler's Zeitschrift fur physiologische Chemie
[Reduction of crotonobetaine and D-carnitine to gamma-butyrobetaine, and the metabolism of L-carnitine in the mouse and rat].
PMID 7409744
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L0399905 Levocarnitine impurity A, European Pharmacopoeia (EP) Reference Standard
C7H13NO2