EMAIL THIS PAGE TO A FRIEND
Journal of virology
Envelope polypeptides of Friend leukemia virus: purification and structural analysis.
PMID 7411688
Abstract
Roughly 10% of surface glycoproteins in the envelope of mature Friend murine leukemia virus are coupled to membrane polypeptides by disulfide bridges. The remaining 90% of these glycoproteins are associated noncovalently. However, they could also be linked to membrane polypeptides by the treatment of purified Friend murine leukemia virus with 2,2'dithiobis(m-nitropyridine). These amphiphilic heterodimer polypeptides, gp84/86, were recovered almost quantitatively in the form of aggregates, termed rosettes, when prepared by solubilization of the viral membrane with Triton X-100 and subsequent velocity sedimentation. gp69/71 and p12(E)/15(E) were purified from these protein micelles after reduction of the disulfide bonds by gel chromatography. Electron micrographs of rosettes, as well as of purified p12(E)/15(E), showed structures different from native viral knobs. Isolated gp84/86 could be reassociated and then displayed more similarity to these viral surface projections. As shown by peptide mapping, the primary structures of the glycoproteins gp69/71 are highly related as are those of the membrane polypeptides p12(E) and p15(E). Furthermore, it was shown by two-dimensional polyacrylamide gel electrophoresis and re-electrophoresis of purified gp84/86 that the larger component, gp86, was composed of gp71 associated with p15(E) and p12(E), whereas the smaller component, gp84, was formed by gp69 bound only to p12(E).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
158194
2,2′-Dithiobis(5-nitropyridine), 96%
C10H6N4O4S2