EMAIL THIS PAGE TO A FRIEND
Doklady Akademii nauk SSSR
[Comitogenic and immunosuppressant action of p,p-DDT and its derivatives on phytohemagglutinin-stimulated human lymphocytes].
PMID 7428595
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
113700
4,4′-Dichlorobenzophenone, 99%
C13H8Cl2O