EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Aerobic and anaerobic degradation of profluralin and trifluralin.
PMID 7430552
Abstract
The degradation of profluralin [N-(cyclopropylmethyl)-alpha,alpha,alpha-trifluoro-2,6-dinitro-N-propyl-p-toluidine] and trifluralin (alpha,alpha,alpha-trifluoro-2,6-dinitro-N,N-dipropyl-p-toluidine) was studied under aerobic and anaerobic soil conditions. Three soils (Goldsboro loamy sand, Cecil loamy sand, Drummer clay loam) were each treated with 1 ppmw herbicide; anaerobic conditions were maintained by flooding. Soil samples were extracted monthly and subjected to TLC analysis. No degradation was detected in sterile controls. Aerobic degradation of both herbicides was greatest in the Cecil loamy sand soil over the entire incubation period. Degradation of profluralin in Cecil soil under aerobic conditions was 86 percent after 4 months with three products detected; 83 percent of the trifluralin was degraded with two products detected. Anaerobic degradation accounted for 72 percent of the profluralin and 78 percent of the trifluralin after 4 months. Degradation of both herbicides increased with incubation time for the first 3 months and decreased slightly thereafter. Generally there was more extensive degradation (percent and in number of products formed) of profluralin than trifluralin under the conditions tested. More degradation products were detected for both herbicides under aerobic conditions than under anaerobic conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45633
Profluralin, PESTANAL®, analytical standard
C14H16F3N3O4