EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
The use of [18O4]phosphoric acid in the quantitation of phosphate by gas-liquid chromatography-mass spectrometry analysis.
PMID 7435965
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
275794
Tris(trimethylsilyl) phosphate, ≥98%
C9H27O4PSi3