EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Dislodgable insecticide residues on cotton foliage: acephate, AC 222,705, EPN, fenvalerate, methomyl, methyl parathion, permethrin, and thiodicarb.
PMID 7437564