EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
Determination of three germicidal phenols in serum of infants with hyperbilirubinemia.
PMID 7438459
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
153842
4-tert-Amylphenol, 99%
C11H16O