EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Determination of maximum permissible concentration of iodofenphos in the water reservoirs].
PMID 7439722
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45544
Jodfenphos, PESTANAL®, analytical standard
C8H8Cl2IO3PS