EMAIL THIS PAGE TO A FRIEND
Kardiologiia
[Antifibrillatory activity of propranolol in experimental myocardial infarct and its cardiodepressive and hypotensive action].
PMID 7441964
Abstract
In experiments in nembutal anesthetized dogs the authors studied the dependence between the antifibrillation activity of propranolol, which was appraised according to the value of the ventricular fibrillation thresholds and the rate of spontaneous fibrillation after occlusion of the coronary artery, and the cardiodepressive and hypotensive action of this agent. It is shown that it is possible in principle to preserve the antifibrillation activity of propranolol infused intravenously in a dose of 1 mg/kg for correcting its cardiodepressive and hypotensive effect in combined intravenous infusion of 0.02 mg/kg corglycon. It is concluded that there is no dependence between antifibrillation activity and the cardiodepressive and hypotensive effects of propranolol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9140
Convallatoxin, ≥65%
C29H42O10