EMAIL THIS PAGE TO A FRIEND
Radiobiologiia
[Effect of xanthobin on the degree of damage to DNA molecules by gamma-irradiation].
PMID 7443994
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
775789
8-Bromocaffeine, 98%
C8H9BrN4O2