EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
Metabolic studies on pentachloronitrobenzene (PCNB) in rats.
PMID 7445521
Abstract
The metabolism of PCNB in rats was studied. Metabolites isolated from rat excreta and identified were: N-acetyl-S-(pentachlorophenyl)cysteine, pentachlorothiophenol, pentachlorothioanisole, 2,3,4,5-tetrachlorothiophenol, 2,3,4,5-tetrachlorothioanisole, 2,3,4,6- and/or 2,3,5,6-tetrachloro-thiophenol and -thioanisole, 1,4-bis(methylthio)tetrachlorobenzene, 1,4-dimercapto-tetrachlorobenzene and/or 4-methylthio-tetrachlorothiophenol, pentachlorophenol, pentachloroanisole, 2,3,4,5-tetrachlorophenol, 2,3,4,5-tetrachloroanisole, 2,3,4,6- and/or 2,3,5,6-tetrachloro-phenol and -anisole, pentachlorobenzene, 2,3,4,5-tetrachloronitrobenzene, pentachloroaniline and 2,3,4,5-tetrachloroaniline.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46012
Pentachloroaniline, PESTANAL®, analytical standard
C6H2Cl5N