EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A 59Co NMR relaxation probe of macromolecule anionic binding sites.
PMID 7447004
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12902
Potassium hexacyanocobaltate(III), ≥97.0%
C6CoK3N6