EMAIL THIS PAGE TO A FRIEND
Archives of toxicology
Repeated small doses of a neurotoxic organophosphate. Monitoring of neurotoxic esterase in brain and spinal cord.
PMID 7447700
Abstract
The effects of small repeated oral doses of mono-2-cresyl diphenyl phosphate (MOCP, 2.5 mg/kg/day) on hen brain and spinal cord neurotoxic esterase (NTE) were measured. The enzyme levels were depressed to about 40% and 55% of normal respectively and maintained at that level for 8 weeks. No clinical and only doubtful histological signs of neuropathy were detected. Neuropathy could be precipitated by depressing the level to < 20% either with a single high dose (50 mg/kg), or by an increase of the repeated dose level to 5 mg/kg/day. There was no correlation between inhibition of NTE in the nervous tissue and the "NTE-like" activity in lymphocytes. "NTE-like" activity in spleen was consistently inhibited but to a lesser extent than that in the brain or spinal cord. Brain AChE and BuChE were not affected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32957
Cresyl diphenyl phosphate, PESTANAL®, analytical standard
C19H17O4P