EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Effects on the cytoskeleton of a new inducer of the neuroblastoma morphological differentiation.
PMID 7447946
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
142824
1-Methyl-1-cyclohexanecarboxylic acid, 99%
C8H14O2