EMAIL THIS PAGE TO A FRIEND
Przeglad dermatologiczny
[Studies on the effect of 3 varieties of an anti-adhesive mineral oil izoform on the skin of mice].
PMID 7455151
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7608
4-Iodoanisole, 98%
C7H7IO