EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[The metabolism of 3H-curcumol in normal rats and tumour bearing mice (author's transl)].
PMID 7457146
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36236
( −)-Curcumol, analytical standard
C15H24O2