EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Spectrophotometric determination of carnosine, anserine, and taurine in skeletal muscle.
PMID 7457874
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
16230
4-Bromoaniline, ≥99.0% (GC)
C6H6BrN
100900
4-Bromoaniline, 97%
C6H6BrN