EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Light-enhanced cross-linking of rhodopsin in rod outer segment membranes as detected by chemical probes.
PMID 7459357
Abstract
Bovine rod outer segment membranes were treated with cross-linking reagents before and after light exposure. Bleached membranes showed enhanced cross-linking with difluorodinitrobenzene or methyl acetimidate compared to dark-adapted membranes. The light-induced enhancement of cross-linking may be due to increased association of rhodopsin monomers in the light and/or due to increased reactivity of amino and sulfhydryl groups of bleached rhodopsin. In some instances, the band ascribed to the rhodopsin monomer in gel electrophoresis appears as a partially resolved doublet. Treatment of bleached rod outer segment membranes with methyl acetimidate improved the resolution of the doublet into two closely migrating bands.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
254940
Methyl acetimidate hydrochloride, technical grade
C3H7NO · HCl