EMAIL THIS PAGE TO A FRIEND
Journal of steroid biochemistry
Synthesis of fluorescein labelled steroid hormone-albumin conjugates for the fluorescent histochemical detection of hormone receptors.
PMID 7464129