EMAIL THIS PAGE TO A FRIEND
Organic Process Research & Development
Advances in Photocatalysis: A Microreview of Visible Light Mediated Ruthenium and Iridium Catalyzed Organic Transformations.
PMID 747793