EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Functional implications for the microtubule-associated protein tau: localization in oligodendrocytes.
PMID 7479786