EMAIL THIS PAGE TO A FRIEND
Planta medica
Cytotoxicity of diterpenes from Premna schimperi and Premna oligotricha.
PMID 7480188
Abstract
The cytotoxicity of two diterpenes from Premna schimperi and Premna oligotricha (Verbenaceae) was studied using the MTT assay. Their cytotoxic activity against three human (HeLa, SK.N.SH, and ECV 304) and two murine (L929 and RAW 264.7) carcinoma cell lines varied between 1.5 to 35 micrograms/ml and was comparable with azauridine and chlorambucil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1882
6-Azauridine
C8H11N3O6