EMAIL THIS PAGE TO A FRIEND
Acta physiologica Scandinavica
Inhibition of leucotriene-synthesis improves dorsal skin flap survival in the rat.
PMID 7484177
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7559
BW A4C, ≥98% (HPLC)
C17H17NO3