EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
Chemotactic peptide analogues. Synthesis and chemotactic activity of N-formyl-Met-Leu-Phe analogues containing (S)-phenylalaninol derivatives.
PMID 7487425
Abstract
The synthesis and the biological activity towards human neutrophils of some N-formyl-Met-Leu-Phe-OMe analogues containing (S)-phenylalaninol (Pheol) or its derivatives in place of the native phenylalanine are reported. While the analogue containing Pheol (4) was found to be devoid of significant biological activity, its esters 3 and 5, although inactive as chemoattractants, are able to strongly stimulate superoxide production and are active with a lower efficacy in the lysozyme release.