EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Synthesis and mass-spectrometric characterization of human serum albumins modified by covalent binding of two non-steroidal anti-inflammatory drugs: tolmetin and zomepirac.
PMID 7487878
Abstract
Human serum albumins modified by covalently bound tolmetin or zomepirac were synthesized as models for similar products formed in vivo from acyl glucuronides. Activated esters of both drugs were prepared with 1-ethyl-3-(3-dimethylaminopropyl)-carbodi-imide, and then allowed to react with human serum albumin. Tryptic digests of both protein products were analysed by HPLC to identify peptides containing covalently bound drugs, and binding sites on albumin were identified by high-performance tandem MS. Three binding sites were common to both products, i.e. lysine-195, -199 and -351. Three further modified residues were identified for the tolmetin-albumin product, i.e. aspartic acid 1, and lysine-524 and -536.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z2625
Zomepirac sodium salt
C15H13ClNNaO3