EMAIL THIS PAGE TO A FRIEND
Diabetologia
Disordered gastric motor function in diabetes mellitus.
PMID 7489851
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5794
GR 79236X, ≥99% (HPLC), solid
C15H21O5N5