EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Chemical constituents of Astragalus chinensis L].
PMID 7492362
Abstract
Three compounds have been isolated from the seeds of Astragalus chinensis. Their structures were identified on the basis of spectral data (UV, 13CNMR, 1HNMR, IR and MS) as octacosane, daucosterol and kaempferol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O504
Octacosane, 99%
C28H58
442696
Octacosane, analytical standard
C28H58