EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
Analysis by limited proteolysis of domain organization and GSH-site arrangement of bacterial glutathione transferase B1-1.
PMID 7496993
Abstract
Limited proteolysis method has been used to study the structure-function relationship of bacterial glutathione transferase (GSTB1-1). In absence of three-dimensional structural data of prokaryote GST, the results represent the first information concerning the G-site and domains organization of GSTB1-1. The tryptic cleavages occur mainly at the peptide bonds Lys35-Lys36 and Phe43-Leu44, generating two major molecular species of 20-kDa, 3-kDa and traces of 10-kDa. 1-chloro-2,4-dinitrobenzene favoured the proteolysis of the 20-kDa fragment markedly enhancing the production of the 10-kDa peptide by cleaving the chemical bonds Lys87-Ala88 and Arg91-Tyr92. The tryptic cleavage sites of GSTB1-1 was found to be located close to those previously found for the mammalian GSTP1-1 isozyme. It was concluded that despite their low sequence homology (18%), GSTB1-1 and GSTP1-1 displayed similar structural features in their G-site regions and probably a common organization in structural domains.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4139
S-Methylglutathione
C11H19N3O6S