EMAIL THIS PAGE TO A FRIEND
FEBS letters
Co-expression of the neurokinin NK2 receptor and G-protein components in the fission yeast Schizosaccharomyces pombe.
PMID 7498495
Abstract
The fission yeast Schizosaccharomyces pombe has proven useful for studying molecular interactions between a range of signal transduction components. We now report the first co-expression of a mammalian seven-transmembrane receptor and G-protein components in S. pombe. We selected the human neurokinin NK2 receptor together with its G-protein-signalling partner Gq for this study. Yeast membrane fractions showed high levels of NK2 receptor-binding activity (1159 +/- 534 (n = 3) fmol/mg protein) although initial experiments with intact cells revealed an absence of receptors at the cell surface. Using a construct comprising the NK2 coding sequence fused with the signal sequence from an endogenous phosphatase (phoI), we detected approximately 400 NK2 receptors/cell in unbroken yeast. Successful co-expression of the NK2 receptor with the G-protein subunits G alpha q, beta 1 or beta 2 and gamma 3 failed to modulate agonist binding, suggesting the absence of functional interaction between these components. As an alternative test of G alpha q function, we next expressed its downstream effector target phospholipase C-beta 1 (PLC beta 1) in S. pombe. Although PLC beta 1 undergoes powerful in vitro activation by G alpha q derived from baculovirus-infected Sf9 cells and mammalian cells, G alpha q expressed in S. pombe is totally ineffective. Similar results were also achieved with the G-protein subunit G alpha 16. Together, these data suggest that seven-transmembrane receptors can be expressed in S. pombe at high levels and directed to the cell surface although their interaction with co-expressed G-proteins in undetectable. Production of inactive G alpha-chains in S. pombe may account for these observations.